2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid ≥99%, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid
BES is a zwitterionic buffer that is used in biochemistry and molecular biology research in the pH range of 6.15 - 8.35 for wide applicability to biochemical studies.
- Presentation: White powder
A protocol has been reported for the use of BES buffered saline in the calcium phosphate mediated transfection of eukaryotic cells with plasmid DNA. BES, a sulfonic acid-containing cross-linking agent, induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes.
The effect of BES and other zwitterionic buffers on the Bicinchoninic acid (BCA) measurement of microgram quantities of protein has been studied. BES was found to diminish color development of the protein BCA complex by 30-35% at 50 mM, and by
BES may be employed as binding buffer in modified Eagle′s medium during the binding assay of human melanoma cells. It may be used as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.
Soluble 0.1M in water (clear, colorless solution)
BES is a zwitterionic buffer that is used in biochemistry and molecular biology research in the pH range of 6.15 - 8.35 for wide applicability to biochemical studies. A protocol has been reported for the use of BES buffered saline in the calcium phosphate mediated transfection of eukaryotic cells with plasmid DNA
Formula:
C₆H₁₅NO₅S MW: 213.25 g/mol Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007533 CAS Number: 10191-18-1 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
pH ( 1% aq soln) | 3 - 5 |
Assay (titration, dry basis) | ≥99.0% |
Solubility (0.1M in H₂O) | Clear, Complete |
Moisture | ≤1.0% |
pKa | 6.9 - 7.3 |
UV A260 nm | ≤0.10 |
UV A280 nm | ≤0.08 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...