Oleoyl coenzyme A dipotassium salt ≥90%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: cis-9-Octadecenoyl coenzyme A potassium salt

0210090401 0210090405 0210090410 0210090450
IC10090401EA 61.33 USD
IC10090401 IC10090405 IC10090410 IC10090450
Oleoyl coenzyme A dipotassium salt ≥90%, Powder
Oleoyl coenzyme A dipotassium salt

Soluble in water.


Coenzyme A functions as an acyl group carrier, acetyl-CoA. Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.

Formula: C₃₉H₆₆N₇O₁₇P₃SK₂·3H₂O
MW: 1162.2 g/mol
MDL Number: MFCD00871328
CAS Number: 1716-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR