DL-Lithium lactate, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt

0210082405 0210082410 0210082450
IC10082405EA 53.52 USD
IC10082405 IC10082410 IC10082450
DL-Lithium lactate, powder
DL-Lithium lactate

Lactic acid is an α hydroxyl carboxylic acid produced from pyruvate by the enzyme lactate dehydrogenase (LDH). Lactate is one of the main components of Ringer′s lactate or lactated Ringer′s solution. It is a fermentation acid from milk or carbohydrates.


Soluble in water.


Precaución: Research Use Only (RUO).

Formula: H₃CCH(OH)COOLi
MW: 96.01
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00065399
CAS Number: 867-55-0

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR