L(+)-Aspartic acid ≥99%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02100809.1 02100809.5 0210080901
IC100809.1EA 65.05 USD
IC100809.1 IC100809.5 IC10080901
L(+)-Aspartic acid ≥99%, Powder
L(+)-Aspartic acid

More soluble in salt solutions; soluble in water acids, alkalies; insoluble in alcohol.


L-Aspartic acid is an endogenous NMDA receptor agonist. Principal neurotransmitter for fast synaptic excitation.

Formula: C₄H₇NO₄
MDL Number: MFCD00002616
CAS Number: 56-84-8

Order Now

Specification Test Results

Synonyms (S)-(+)-Aminosuccinic acid; (S)-Aminobutanedioic acid
Form White Powder
Storage Store at Room Temperature(15-30 °C).
Loss on Drying ≤0.5%
Optical Rotation +21 to +27° (c=1, 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR