Guanosine 5'-triphosphate disodium salt ≥98%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt
Guanosine-5'-triphosphate, a purine nucleotide is required as a coenzyme for protein biosynthesis in a cell-free system.
Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF).
GTP functions as a carrier of phosphates and pyrophosphates involved in channeling chemical energy into specific biosynthetic pathways. GTP activates the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades. Proliferation and apoptosis are regulated in part by the hydrolysis of GTP by small GTPases Ras and Rho. Another type of small GTPase, Rab, plays a role in the docking and fusion of vesicles and may also be involved in vesicle formation. In addition to its role in signal transduction, GTP also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.
Soluble in water (50 mg/mL - clear and colorless).
Guanosine-5'-triphosphate, a purine nucleotide is required as a coenzyme for protein biosynthesis in a cell-free system.
Precaución: Research Use Only (RUO).
Formula:
C₁₀H₁₄N₅Na₂O₁₄P₃ MW: 567.147 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03410297 CAS Number: 56001-37-7 |
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
UV/VIS: λmax (water) | 252 ± 3 nm |
OD250/OD260 | 1.20 ± 0.08 |
OD280/OD260 | 0.65 ± 0.08 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...