Dansyl chloride ~98%, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

0210060301 0210060305
IC10060301EA 141.13 USD
IC10060301 IC10060305
Dansyl chloride ~98%, yellow powder
Dansyl chloride

Dansyl Chloride is a fluorogenic reagent for fluorescent N-terminal derivatization of amino acids and peptides and detection by reverse phase HPLC. It is used in labeling proteins and amino acids.


Soluble in acetone (10% w/v - clear solution).


Dansyl Chloride is a fluorogenic reagent for fluorescent N-terminal derivatization of amino acids and peptides and detection by reverse phase HPLC. It is used in labeling proteins and amino acids.

Formula: C₁₂H₁₂ClNO₂S
MW: 269.75
Melting Pt: 64…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
UN: 3261
ADR: 8 III
Merck Index: 13,02840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR