DL-α-Tocopherol acetate, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210055905 0210055925 0210055980
IC10055905EA 45.7 USD
IC10055905 IC10055925 IC10055980
DL-α-Tocopherol acetate, liquid
DL-α-Tocopherol acetate

Tocopherol acetate has properties similar but not identical to α-tocopherol.


Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens.


Freely soluble in absolute ethanol, acetone, chloroform (100 mg/mL-clear solution), ether and fixed oils, soluble in ethanol (96%); slightly soluble in alcohol; practically insoluble in water.

Formula: C₃₁H₅₂O₃
MDL Number: MFCD00072042
CAS Number: 7695-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR