Creatine phosphate disodium salt tetrahydrate ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

0210052001 0210052005 0210052025 02100520.2
IC10052001EA 104.08 USD
IC10052001 IC10052005 IC10052025 IC02100520.2
Creatine phosphate disodium salt tetrahydrate ≥98%, white powder
Creatine phosphate disodium salt tetrahydrate

Creatine phosphate is substrate for CPK determination.


  • Substantially free of creatine


Soluble in water (150 mg/ml-clear, colorless solution)

Formula: C₄H₈N₃Na₂O₅P·4H₂O
MW: 327.14 g/mol (255.07 anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150192
CAS Number: 71519-72-7

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) 7 - 9
Moisture 22 ± 3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR