β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) ≥98%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized

02100499.1 02100499.5
IC100499.1EA 41.93 USD
IC100499.1 IC100499.5
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) ≥98%
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidized form)

β-NAD is one of the biologically active forms of nicotinic acid. It occurs in living cells primarily in the oxidized state. Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols. NAD usually acts as a hydrogen acceptor, forming NADH which then serves as a hydrogen donor in the respiratory chain. (Merck Index, 12th Ed., No.6429)


  • White powder, soluble in water (200 mg/ml)
  • pH: 2 - 4 (1% aqueous solution)


β-nicotinamide adenine dinucleotide is a major electron acceptor molecule in biological oxidations. It acts as a coenzyme for hydride-transfer enzymes and a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins. Recently reported to be a P2Y11 agonist.


Solutions should be aliquoted and stored at –20 °C for up to one week.

Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
MDL Number: MFCD00036253
CAS Number: 53-84-9

Order Now

Specification Test Results

Form White Powder
Storage Store at - 0 °C. Store Desiccated.
pH 2 - 4 (1% aquous solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR