Coenzyme A trilithium salt dihydrate ≥96%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Ubidecarenone (coenzyme, q-10), CoA Li3, Trilithium coenzyme A

0210049325 0210049350 0210049380 0210049383 0210049390 0210049391 0210049310
IC10049325EA 134.01 USD
IC10049325 IC10049350 IC10049380 IC10049383 IC10049390 IC10049391 75875-622
Coenzyme A trilithium salt dihydrate ≥96%, white crystalline powder
Coenzyme A trilithium salt dihydrate

Coenzyme A is an essential cofactor in enzymatic acetyl transfer reactions. The principal biologically active forms of pantothenic acid are coenzyme A (CoA) and acyl carrier protein (ACP). In CoA, the business center of the molecule is the pantothenic acid metabolite 4'-phosphopantetheine. Coenzyme A is comprised of 4'-phosphopantetheine linked by an anhydride bond to the nucloetide adenosine 5'-monophosphate. 4'-Phosphopantetheine itself is comprised of pantothenic acid linked at one end, via an amide bond, to beta-mercaptoethylamine, derived from L-cysteine, and at the other end to a phosphate group. The sulfhydryl group of 4'-phosphopantetheine, which is the business end of the coenzyme, forms thioesters with acyl groups producing acyl-CoA derivatives, including acetyl-CoA.
Coenzyme A facilitates removal of lipid peroxides by increasing mobilization of fatty acids, and promote repair of plasma membranes by activating phospholipid synthesis.


  • Presentation: White Crystalline Powder
  • Soluble in water (50 mg/mL - clear, colorless to faint yellow solution)
  • Storage temperature: -20 °C

Formula: C₂₁H₃₃N₇O₁₆P₃SLi₃·2H₂O
MW: 821.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00075848
CAS Number: 18439-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=330bd3624b0fa28c784869156e8963db921e5aab&vid=2284d06837ee1651dcfbe0fcca4992c56788d8bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥96%
OD250/OD260 (pH 2) 0.80-0.89

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR