Spermidine trihydrochloride ≥99%, crystalline

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210047201 0210047205
IC10047201EA 65.46 USD
IC10047201 IC10047205
Spermidine trihydrochloride ≥99%, crystalline
Spermidine trihydrochloride

Spermidine is a biogenic polyamine formed from putrescine. It is essential in both normal and neoplastic tissue growth. It plays an important role in the regulation of cellular proliferation and differentiation. Forms stable compounds with nucleic acids. Inhibits neuronal nitric oxide synthase (nNOS).


Spermidine binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity. It has a role in cell growth processes. It can be used in electroporation while transferring the DNA into the cell under the electrical impulse. It is also plant growth regulator, promoting somatic embryogenesis.


Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.


Soluble in water (100 mg/mL - clear, colorless solution). MP Biomedicals recommends solutions should be made fresh for each use; however, if this is not possible, aliquot solutions and store at -20 °C or below for short periods.


Spermidine is a biogenic polyamine formed from putrescine. It is essential in both normal and neoplastic tissue growth. It plays an important role in the regulation of cellular proliferation and differentiation. Forms stable compounds with nucleic acids. Inhibits neuronal nitric oxide synthase (nNOS).


Precaución: Research Use Only (RUO).

Formula: C₇H₂₂Cl₃N₃
Storage Temperature: Freezer
MDL Number: MFCD00012918
CAS Number: 334-50-9

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 253 - 263 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR