1,4-Diaminobutane dihydrochloride ≥98%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0210045001 0210045025 0210045080
IC10045001EA 42.83 USD
IC10045001 IC10045025 IC10045080
1,4-Diaminobutane dihydrochloride ≥98%, crystalline powder
1,4-Diaminobutane dihydrochloride

Putrescine is a biogenic polyamine and precursor of spermidine. It is formed via decarboxylation of ornithine or by decarboxylation of arginine followed by hydrolysis.


Binds to the polyamine modulatory site of the NMDA receptor and potentiates NMDA-induced currents.


Presentation: White Crystalline Powder


Soluble in water (10 mg/mL - clear, colorless solution).


Precaución: Store at room temperature (15 - 30 °C). Store Desiccated. Protect from light.

Formula: C₄H₁₄Cl₂N₂
Melting Pt: 280 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00012526
CAS Number: 333-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b18abcc2e044428458b9314572cac399a8766105&vid=b18abcc2e044428458b9314572cac399a8766105&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
pH (1% aq soln) 6 - 7
Residue on Ignition 0.1%
Heavy Metals ≤20ppm
Loss on Drying 0.18%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR