Nitro blue tetrazolium chloride (NBT), yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

0210041650 0210041683 0210041690 0210041691 0210041694
IC10041650EA 50.95 USD
IC10041650 IC10041683 IC10041690 IC10041691 IC10041694
Nitro blue tetrazolium chloride (NBT), yellow powder
Nitro blue tetrazolium chloride (NBT)

p-Nitroblue Tetrazolium Chloride is a substrate for dehydrogenases and other oxidases and precipitating agent for fibrinogen.


p - nitro blue tetrazolium is used for the detection of alkaline phosphatase on western blots. It has also been used as a redox indicator for other enzymatic reactions including dehydrogenases, threonine deaminase, glucose-6-phosphate dehydrogenase, phosphofructokinase on polyacrylamide gels, oxidases on polyacrylamide gels, and pentose shunt dehydrogenases. Redox and halfwave potentials have been determined for p - nitro blue tetrazolium. It has also been used as a colorimetric indicator of bacterial infection in blood samples.


Slightly (10 mg/mL) soluble in water.


p-Nitroblue Tetrazolium Chloride is a substrate for dehydrogenases and other oxidases and precipitating agent for fibrinogen.

Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 189…192 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9

Order Now

Specification Test Results

identity Test Passes
l max (MeOH) 255 ± 3 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR