Alizarin red S (sodium salt), anhydrous, orange powder C.I. 58005
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium
0210037525
0210037580
0210037590
IC10037525EA
83.72
USD
IC10037525
IC10037580
IC10037590
Alizarin red S (sodium salt), anhydrous, orange powder C.I. 58005
Alizarin Red S (sodium salt)
Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.
Soluble in water (1 mg/ml).
Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.
Formula:
C₁₄H₇NaO₇S Storage Temperature: Ambient |
MDL Number:
MFCD00013049 CAS Number: 130-22-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...