5-Iodo-2'-deoxyuridine (Idoxuridine) ∼99% (by HPLC)

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02100357.2 0210035701 0210035705 0210035725
IC100357.2EA 45.75 USD
IC100357.2 IC10035701 IC10035705 IC10035725
5-Iodo-2'-deoxyuridine (Idoxuridine) ∼99% (by HPLC)
5-Iodo-2'-deoxyuridine (Idoxuridine)

5-Iodo-2′-deoxyuridine is pyrimidine analog (halogenated thymidine) compete with thymidine for incorporation into the DNA of replicating cells. This incorporation results in radio sensitizing effects which is used in human cancer studies .


5-Iodo-2'-deoxyuridine (5-IUdR; IdUrd) is a pyrimidine analog (halogenated thymidine) that has antiviral activity agains vaccinia virus (orthopoxviruses). IdUrd is used as a radiosensitizer in human cancer studies.


Solubility at 25 °C in mg/mL: 2.0 in water; 2.0 in 0.2N HCl; 74.0 in 0.2N NaOH; 4.4 in methanol; 2.6 in alc; 0.014 in ether; 0.003 in chloroform; 1.6 in acetone; 1.8 in ethyl acetate; 5.7 in dioxane.


5-Iodo-2′-deoxyuridine is pyrimidine analog (halogenated thymidine) compete with thymidine for incorporation into the DNA of replicating cells. This incorporation results in radio sensitizing effects which is used in human cancer studies .

Formula: C₉H₁₁IN₂O₅
MDL Number: MFCD00134656
CAS Number: 54-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18c48ca579eb0f0aab08cc5b6da1b6eff2304681&vid=18c48ca579eb0f0aab08cc5b6da1b6eff2304681&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR