Histamine diphosphate, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate

0210034301 0210034305 0210034325
IC10034301EA 48.55 USD
IC10034301 IC10034305 IC10034325
Histamine diphosphate, crystalline powder
Histamine diphosphate

Histamine Diphosphate Monohydrate is potent vasodilater.


Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2â‚‚ activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.


Soluble in water (50 mg/mL - clear, colorless to almost colorless solution).


Histamine Diphosphate Monohydrate is potent vasodilater.


Precaución: Research Use Only (RUO).

Formula: C₅H₉N₃·2H₃PO₄
MW: 307.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00064316
CAS Number: 51-74-1

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Loss on Drying Report result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR