D-Glucose-6-phosphate dipotassium salt hydrate ≥95%, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
02100313.5
0210031301
0210031305
IC100313.5EA
89.83
USD
IC100313.5
IC10031301
IC10031305
D-Glucose-6-phosphate dipotassium salt hydrate ≥95%, powder
D-Glucose-6-phosphate dipotassium salt hydrate
Soluble in water (50 mg/mL-clear, colorless).
Formula:
C₆H₁₁K₂O₉P MW: 336.315 g/mol |
CAS Number:
5996-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...