1-Naphthyl phosphate disodium salt monohydrate, Crystalline

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 1-Naphthyl phosphate, disodium salt hydrate

02100257.1 02100257.2 0210025701 0210025705
IC100257.1EA 41.64 USD
IC100257.1 IC100257.2 IC10025701 IC10025705
1-Naphthyl phosphate disodium salt monohydrate, Crystalline
1-Naphthyl phosphate disodium salt monohydrate

α - Naphthyl phosphate disodium salt is a substrate for determination of phosphatase.


1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.


Soluble in water (50 mg/ml)


α - Naphthyl phosphate disodium salt is a substrate for determination of phosphatase.

Formula: C₁₀H₇Na₂O₄P·1H₂O
MW: 268.12 g/mol (anhy)
Melting Pt: 157…159 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00150614
CAS Number: 207569-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27036284e383289c65fb531364e83a393b426f26&vid=27036284e383289c65fb531364e83a393b426f26&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR