Adenine sulfate dihydrate ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 6-Aminopurine sulphate dihydrate, Adenine hemisulphate dihydrate, 7H-Purin-6-amine hemisulphate dihydrate, 9H-Purin-6-amine hemisulphate dihydrate

0210019501 0210019525 0210019505 0210019580
IC10019501EA 60.29 USD
IC10019501 IC10019525 IC0210019505 IC0210019580
Adenine sulfate dihydrate ≥99%, powder
Adenine sulphate dihydrate

Adenine is a purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro. It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).


Solubility (0.5% aq soln): Clear, Colorless Solution.


Adenine is a purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro.It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).

Formula: (C₅H₅N₅)₂·H₂SO₄·2H₂O
MW: 404.37 g/mol
Melting Pt: 260 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS Number: 6509-19-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae66659308f501322c725c1f448f1ffd4ff2b0b5&vid=ae66659308f501322c725c1f448f1ffd4ff2b0b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity 101.7% (UV)
FTIR Conforms to Standard
Moisture 8,60%
pH (0.5% aq soln) 3,04
UV/VIS: l max (water) 260.6 nm
OD250/OD260 0.78
OD280/OD260 0.113
Solubility (0.5% aq soln) Clear, Colorless Solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR