Adenine hydrochloride ~ 99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride

0210019105 0210019125 0210019180
IC10019105EA 71.54 USD
IC10019105 IC10019125 IC10019180
Adenine hydrochloride ~ 99%, white crystalline powder
Adenine hydrochloride

Adenine is a purine nucleobase. It is widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids; a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase.


Adenine has a wide range of chemical and biochemical roles in vivo and in vitro. It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP). It is used as a blood preservative and in viral research.


Soluble in water (1 g/42 ml).Soluble in 50% ammonium hydroxide (30 mg/ml - clear, colorless solution).


Adenine is a purine nucleobase. It is widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids; a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase.


Precaución: Research Use Only (RUO).

Formula: C₅H₅N₅·HCl
MW: 171.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150864
CAS Number: 2922-28-3

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
UV/VIS: λ max (0.1N HCl) 261 - 263 nm
OD250/OD260 0.75 - 0.85
OD280/OD260 0.25 - 0.35
OD290/OD260 0.025 - 0.035
Heavy Metals ≤10 ppm
Sulphated Ash ≤0,1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR