N-Acetyl-β-D-mannosamine hydrate ~95%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02100109.2 0210010901 0210010905
IC100109.2EA 47.08 USD
IC100109.2 IC10010901 IC10010905
N-Acetyl-β-D-mannosamine hydrate ~95%, white powder
N-Acetyl-β-D-mannosamine hydrate

N-Acetyl-β-D-mannosamine hydrate is used as a substrate to identify, differentiate and characterise enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s). N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.


N-Acetyl-D-mannosamine (ManNAc) is an essential precursor of N-acetylneuraminic acid (NeuAc), the specific monomer of polysialic acid. Transport studies in the E. coli K1 strain have been performed to probe the effect of ManNAc on capsular polysialic acid production.
Soluble in water (50 mg/ml - clear, colorless solution).


Chloride ≤50 ppm


N-Acetyl-β-D-mannosamine hydrate is used as a substrate to identify, differentiate and characterize enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s). N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.


Precaución: Research Use Only (RUO).

Formula: C₈H₁₅NO₆·H₂O
MW: 239.2 g/mol
Storage Temperature: Freezer
CAS Number: 4773-29-9

Order Now

Specification Test Results

Identity Test Passes
Purity ~95%
FTIR Conforms to Standard
Specific Rotation +10,2° ± 0,5°
Moisture Content ≤8,0%
Residue on Ignition ≤0,1%
Heavy Metals ≤10 ppm
Chloride (Cl) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR