N-Acetyl-β-D-glucosamine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

0210006825 0210006880 0210006810
IC10006825EA 75.33 USD
IC10006825 IC10006880 IC0210006810
N-Acetyl-β-D-glucosamine, white powder
N-Acetyl-β-D-glucosamine

N-Acetyl-D-glucosamine (D-GlcNAc) is a derivitized glucose monomer found in polymers of bacterial cell walls, chitin, hyaluronic acids and various glycans.


N-Acetyl-D-glucosamine is used to identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s).


Solubility: Soluble in water (50 mg/ml).


N-Acetyl-D-glucosamine (D-GlcNAc) is a derivitized glucose monomer found in polymers of bacterial cell walls, chitin, hyaluronic acids and various glycans.


Precaución: Research Use Only (RUO).

Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Melting Pt: 199 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00136044
CAS Number: 7512-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ab3f10646ca4262bc4d5ac0af6991aa01da81c2&vid=7ab3f10646ca4262bc4d5ac0af6991aa01da81c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Loss on Drying 0,57%
Optical Rotation +43.4°
Transmittance 96,9%
Chloride 0,0005
Heavy Metals <10 ppm
Arsenic <1 ppm
Residue on Ignition 0,0001
pH 7.2
Total Plate Count ≤500 cfu/g
Mold & Yeast ≤100 cfu/g
E.coli, Samonella, Staphylococcus aureus, & Coliforms Negative

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR