N-Acetyl-D-galactosamine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02100064.1 02100064.5 0210006401 0210006405 0210006425
IC100064.1EA 60.74 USD
IC100064.1 IC100064.5 IC10006401 IC10006405 IC10006425
N-Acetyl-D-galactosamine, white powder
N-Acetyl-D-galactosamine

Soluble in water (100 mg/mL - clear, colorless solution).


N-Acetyl-D-galactosamine(GalNAc), an aminosugar, is a component of many O-linked and N-linked glycan structures.


N-Acetyl-D-galactosamine is the intial O-linked sugar to many serine and threonine residues in protein glycosylations.


Soluble in water (100 mg/mL - clear, colorless solution).


N-Acetyl-D-galactosamine(GalNAc), an aminosugar, is a component of many O-linked and N-linked glycan structures.

Formula: C₈H₁₅NO₆
Melting Pt: 160…161 °C (lit.)
MDL Number: MFCD00065372
CAS Number: 1811-31-0

Order Now

Specification Test Results

Synonyms N-Acetylchondrosamine; 2-Acetamido-2-deoxy-D-galactose; D-GalNAc
Form White Powder
Storage Store at +4 °C.
Moisture Content <5.0%
Optical Rotation +84 ° to +89 ° (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR