Adenosine 5'-diphosphate disodium salt (ADP disodium salt) ≥95%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate

02100056.5 0210005601 0210005605
IC100056.5EA 78.54 USD
IC100056.5 IC10005601 IC10005605
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) ≥95%, powder
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)

ATP ≤1%


Adenosine 5′-diphosphate (ADP) is an adenine nucleotide involved in energy storage and nucleic acid metabolism via its conversion into ATP by ATP synthases. ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12 and P2X1. Upon its conversion to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.

Formula: C₁₀H₁₃N₅Na₂O₁₀P₂
MW: 471.167 g/mol
MDL Number: MFCD00066635
CAS Number: 16178-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb2542ee78995af984a49d5e68f818c910e75b4&vid=479e9dedd818e91068cfbf9694c3bd887d5b9e18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms ADP-Na2; ADP
Form White Powder
Storage Store at -20 °C.
Heavy Metals ≤0.002%
Optical Rotation [α]20/D -29.6±1°, c = 0.5% in 0.5 M Na2HPO4(Lit.)
Source Yeast


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR