Acetylcholine chloride ∼99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: (2-Acetoxyethyl)trimethylammonium chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, 2-Acetoxy-N,N,N-trimethylethanaminium chloride

0210001225 0210001280 0210001290
IC10001225EA 111.59 USD
IC10001225 IC10001280 IC10001290
Acetylcholine chloride ∼99%, powder
Acetylcholine chloride

Acetylcholine chloride is a cholinergic neurotransmitter that can induce the opening of calcium channels, as well as act on nicotinic and muscarinic acetylcholine receptors.


Acetylcholine chloride have ophthalmological uses and can be broken down quickly by choliesterases. Studies show that non-neuronal acetylcholine influences many basic cells functions, such as mitosis, cells differentiation, cytoskeletal organization, and cell to cell contact, among other functions.


Acetylcholine chloride, injected at 20 mg/kg body weight, reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis . The cholinergicanti-inflammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.


Soluble in water (100 mg/mL), alcohol; insoluble in ether.


Acetylcholine chloride is a cholinergic neurotransmitter that can induce the opening of calcium channels, as well as act on nicotinic and muscarinic acetylcholine receptors.

Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
MW: 181.66 g/mol
Melting Pt: 151
Storage Temperature: Freezer
MDL Number: MFCD00011698
CAS Number: 60-31-1
Merck Index: 13,00089

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 147 - 157 °C
pH (1% aq soln) 4 - 6
Moisture Content ≤1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR