Novobiocin sodium salt 20 mg/ml in distilled water, sterile filtered
Supplier: Thermo Scientific Chemicals
J67381-AD
J67381-AE
AAJ67381-ADEA
246
USD
AAJ67381-AD
AAJ67381-AE
Novobiocin sodium salt 20 mg/ml in distilled water, sterile filtered
Novobiocin sodium salt
Concentration: 20 mg/ml
CAS No.: 1476-53-5
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00066541
Formula:
C₃₁H₃₅N₂NaO₁₁ Storage Temperature: Refrigerator |
MDL Number:
MFCD00066541 CAS Number: 1476-53-5 EINECS: 216-023-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...