2-Methyl-5-nitroimidazole-1-ethanol 50 mg/ml in hydrochloric acid 0.1M, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Metronidazole
J67362-AC
J67362-AD
AAJ67362-ACEA
114
USD
AAJ67362-AC
AAJ67362-AD
2-Methyl-5-nitroimidazole-1-ethanol 50 mg/ml in hydrochloric acid 0.1M, sterile filtered
2-Methyl-5-nitroimidazole-1-ethanol
Metronidazole, 50 mg/ml in 0.1N HCl, sterile-filtered
CAS No.: 443-48-1
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00009750
Formula:
C₆H₉N₃O₃ MW: 171.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00009750 CAS Number: 443-48-1 EINECS: 207-136-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...