Tobramycin sulfate 50 mg/mL in distilled water, sterile-filtered
Supplier: Thermo Scientific Chemicals
J67340-8EQ
J67340-XF
AAJ67340-8EQEA
504
USD
AAJ67340-8EQ
AAJ67340-XF
Tobramycin sulfate 50 mg/mL in distilled water, sterile-filtered
Tobramycin sulfate
Tobramycin sulfate, 50 mg/ml in distilled water, sterile-filtered
- Liquid
CAS No.: 79645-27-5
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00133864
Precaución: Research use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
(C₁₈H₃₇N₅O₉)₂·5H₂SO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00133864 CAS Number: 79645-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...