Piperacillin sodium salt 50 mg/ml in distilled water, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
Piperacillin sodium salt, 50 mg/ml in distilled water, sterile-filtered
CAS No.: 59703-84-3
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00917471
Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₂₃H₂₇N₅O₇S Storage Temperature: Refrigerator |
MDL Number:
MFCD00917471 CAS Number: 59703-84-3 EINECS: 261-868-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...