Trimethoprim 50 mg/mL in DMSO, sterile filtered

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine

J67175-8EQ J67175-XF
AAJ67175-8EQEA 549.97 USD
AAJ67175-8EQ AAJ67175-XF
Trimethoprim 50 mg/mL in DMSO, sterile filtered
Trimethoprim

Trimethoprim, 50 mg/ml in DMSO, sterile-filtered


CAS No.: 738-70-5 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD00036761


Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₁₄H₁₈N₄O₃
MW: 290.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036761
CAS Number: 738-70-5
EINECS: 212-006-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR