Ceftriaxone disodium salt hemiheptahydrate 50 mg/ml in distilled water

Supplier: Thermo Scientific Chemicals
Danger

J67171-8EQ J67171-XF
AAJ67171-8EQEA 356 USD
AAJ67171-8EQ AAJ67171-XF
Ceftriaxone disodium salt hemiheptahydrate 50 mg/ml in distilled water
Ceftriaxone disodium salt hemiheptahydrate

Ceftriaxone disodium salt, 50 mg/ml in distilled water


CAS No.: 104376-79-6 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD01750405

Formula: C₁₈H₁₈N₈O₇S₃
Storage Temperature: Refrigerator
MDL Number: MFCD01750405
CAS Number: 104376-79-6
EINECS: 277-930-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR