Doxycycline hydrochloride 20 mg/mL in distilled water, sterile filtered
Supplier: Thermo Scientific Chemicals
Synonyms:
(4S,4aR,5S,5aR,6R,12aS)-4-(Dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
J67043-AD
J67043-AE
AAJ67043-ADEA
266.49
USD
AAJ67043-AD
AAJ67043-AE
Doxycycline hydrochloride 20 mg/mL in distilled water, sterile filtered
Doxycycline hydrochloride
Doxycycline hydrochloride, 20 mg/ml in distilled water, sterile-filtered
CAS No.: 10592-13-9
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD03427564
Formula:
C₂₂H₂₄N₂O₁₀·HCl MW: 480.9 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03427564 CAS Number: 10592-13-9 EINECS: 234-198-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...