Neomycin sulfate 50 mg/ml in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

J67011-AD J67011-AE
AAJ67011-ADEA 183.3 USD
AAJ67011-AD AAJ67011-AE
Neomycin sulfate 50 mg/ml in distilled water, sterile filtered
Neomycin sulphate

Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered


CAS No.: 1405-10-3 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD00150827

Formula: C₂₃H₄₆N₆O₁₃·3H₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00150827
CAS Number: 1405-10-3
EINECS: 215-773-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR