Cefuroxime sodium salt 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Cefuroxime sodium salt, 95%
Purity: 95%
CAS No.: 56238-63-2
Molecular Formula: C16H15N4NaO8S
Formula Weight: 446.37
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 240-245°
Solubility: Freely soluble in water
Optical Rotation: +60° (c=1 in water)
MDL No.: MFCD09878727
Formula:
C₁₆H₁₅N₄NaO₈S Melting Pt: 240…245 °C Storage Temperature: Ambient |
MDL Number:
MFCD09878727 CAS Number: 56238-63-2 EINECS: 260-073-1 Merck Index: 14,1954 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...