Hyaluronic acid sodium salt (from Streptococcus equi) 91%, powder

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium hyaluronate, Hyaluronic acid sodium, HA sodium salt

J66993-03 J66993-ME
10067-436EA 132.76 USD
10067-436 AAJ66993-ME
Hyaluronic acid sodium salt (from Streptococcus equi) 91%, powder
Hyaluronic acid sodium salt

Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.


Molecular weight is greater than or equal to 1MDa


Applications: An anionic, nonsulfated glycosaminoglycan distributed widely throughout connective, epithelial, and neural tissues


Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: (C₁₄H₂₁NaNO₁₁)n
Storage Temperature: Refrigerator
MDL Number: MFCD01773053
CAS Number: 9067-32-7
Merck Index: 14,4757

Order Now

Specification Test Results

Assay 91%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR