Levofloxacin 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid

J66943-03 J66943-06
10067-432EA 57.92 USD
10067-432 10067-434
Levofloxacin 98%
Levofloxacin

Levofloxacin, 98%


Purity: 98% CAS No.: 100986-85-4 Molecular Formula: C18H20FN3O4 Formula Weight: 361.37 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to pale yellow Melting Point: 218-227° Boiling Point: 572° Solubility: Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane Beilstein Reference: 7327015 MDL No.: MFCD00865049

Formula: C₁₈H₂₀FN₃O₄
MW: 361.37 g/mol
Boiling Pt: 572 °C
Melting Pt: 218…227 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00865049
CAS Number: 100986-85-4
Merck Index: 14,6771

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR