L-Selenomethionine
Supplier: Thermo Scientific Chemicals
|
Danger
|
Seleno-L-methionine
CAS No.: 3211-76-5
Molecular Formula: C5H11NO2Se
Formula Weight: 196.11
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 265°
UN No.: 3283
MDL No.: MFCD00037210
Formula:
C₅H₁₁NO₂Se Melting Pt: 265 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037210 CAS Number: 3211-76-5 UN: 3283 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...