Oleoyl-L-α-lysophosphatidic acid sodium salt
Supplier: Thermo Scientific Chemicals
J66836-LB0
J66836-MB
AAJ66836-LB0EA
178.75
USD
AAJ66836-LB0
AAJ66836-MB
Oleoyl-L-α-lysophosphatidic acid sodium salt
Oleoyl-L-alpha-lysophosphatidic acid sodium salt
Oleoyl-L-^a-lysophosphatidic acid sodium salt
CAS No.: 22556-62-3
Molecular Formula: C21H40NaO7P
Formula Weight: 458.5
Storage Temperature: -30°C to -10°C
Physical Form: Waxy solid or powder
Sensitivity: Light and Air Sensitive; Hygroscopic
Appearance: White to off-white
Solubility: Soluble in water up to 5mg/ml with sonication
MDL No.: MFCD00133427
Formula:
C₂₁H₄₀NaO₇P MW: 458.508 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00133427 CAS Number: 22556-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...