Ascomycin 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone
An analog of tacrolimus (FK-506). Ascomycin is an inhibitor of PP2 and it is potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases
- White powder
It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus.
Soluble in DMSO at 65 mg/mL; Soluble in Ethanol at 50 mg/m; with Warming. Very Poorly Soluble in Water
Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₄₃H₆₉NO₁₂ MW: 790 g/mol Boiling Pt: 868 °C Melting Pt: 156…160 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Freezer |
MDL Number:
MFCD06198665 CAS Number: 104987-12-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...