Imipenem monohydrate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (5R,6S)-3-((2-Formimidamidoethyl)thio)-6-((R)-1-hydroxyethyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate

J66749-LB0 J66749-MB
10067-484EA 114 USD
10067-484 AAJ66749-MB
Imipenem monohydrate 95%
Imipenem monohydrate

A broad spectrum antibiotic


Applications: A broad spectrum antibiotic


Applications: A broad spectrum antibiotic

Formula: C₁₂H₁₇N₃O₄S
MW: 317.36 g/mol
Boiling Pt: 567 °C
Melting Pt: 193…198 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD09753321
CAS Number: 74431-23-5
EINECS: 264-734-5
Merck Index: 14,4919

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR