Sulfamonomethoxine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-N-(6-methoxypyrimidin-4-yl)benzenesulphonamide, 4-Amino-N-(6-methoxy-4-pyrimidinyl)benzenesulphonamide, N-(6-Methoxy-4-pyrimidinyl)sulphanilamide

J66733-03 J66733-MD
10067-264EA 388 USD
10067-264 10067-480
Sulfamonomethoxine 98%
Sulfamonomethoxine

A bacteriostatic sulfonamide antibiotic which acts as a folic acid synthesis inhibitor


Applications: A bacteriostatic sulfonamide antibiotic which acts as a folic acid synthesis inhibitor


Applications: A bacteriostatic sulfonamide antibiotic which acts as a folic acid synthesis inhibitor

Formula: C₁₁H₁₂N₄O₃S
MW: 280.30 g/mol
Boiling Pt: 513 °C
Melting Pt: 203…206 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063466
CAS Number: 1220-83-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR