Fusidate sodium

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate

J66703-03 J66703-06
10067-260EA 260.63 USD
10067-260 10067-262
Fusidate sodium
Fusidate sodium

Targets Gram-positive bacteria namely those which cause skin infections


CAS No.: 751-94-0 Molecular Formula: C31H47NaO6 Formula Weight: 538.69 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: White to off-white Solubility: Soluble in water MDL No.: MFCD09054714


Applications: Targets gram positive bacteria namely those which cause skin infections

Formula: C₃₁H₄₇NaO₆
MW: 538.701 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054714
CAS Number: 751-94-0
EINECS: 212-030-3
Merck Index: 14,4317

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR