Picloram 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-3,5,6-trichloropicolinic Acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic Acid

J66702-06 J66702-14
10067-256EA 161.88 USD
10067-256 10067-258
Picloram 95%
Picloram

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)


Applications: Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)


Applications: Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)

Formula: C₆H₃Cl₃N₂O₂
MW: 241.45 g/mol
Boiling Pt: 421 °C
Melting Pt: 219 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012101
CAS Number: 1918-02-1
EINECS: 217-636-1
Merck Index: 14,7397

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR