Colimycin M

Supplier: Thermo Scientific Chemicals

Synonyms: Polymyxin E sodium methanesulphonate, Colistin sodium methanesulphonate

J66698-03 J66698-06
10067-252EA 177 USD
10067-252 10067-254
Colimycin M
Colimycin M

Has a bactericidal effect by targeting the cell membrane and modifying its permeability


CAS No.: 8068-28-8 Molecular Formula: C58H115N16Na5O28S5 Formula Weight: 1759.89 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Solubility: Freely soluble in water MDL No.: MFCD00130824


Applications: Has a bactericidal effect by targeting the cell membrane and modifying its permeability

Formula: C₅₈H₁₀₅N₁₆Na₅O₂₈S₅
MW: 1749.811 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130824
CAS Number: 8068-28-8
EINECS: 232-516-9
Merck Index: 14,2479

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR