Gibberellic acid 90% A4+A7
Supplier: Thermo Scientific Chemicals
Synonyms:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
J66661-03
J66661-06
10067-238EA
106.13
USD
10067-238
10067-240
AAJ66661-MD
Gibberellic acid 90% A4+A7
Gibberellic acid
Purity: 90%
Storage Temperature: -30°C to -10°C
Flash Point:
Formula:
C₁₉H₂₂O₆ MW: 346.38 g/mol Boiling Pt: 582 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...