5Z-7-Oxozeaenol

Supplier: Thermo Scientific Chemicals
Danger

J66651-LB0 J66651-MCR
AAJ66651-LB0EA 610 USD
AAJ66651-LB0 AAJ66651-MCR
5Z-7-Oxozeaenol
5Z-7-Oxozeaenol

5Z-7-Oxozeaenol


CAS: 66018-38-0 Form: Solid MDL: MFCD12912700

Formula: C₁₉H₂₂O₇
MW: 362.378 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12912700
CAS Number: 66018-38-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR