TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), liquid for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
J66624-0F
J66624-AE
J66624-AP
AAJ66624-0FEA
154.1
USD
AAJ66624-0F
AAJ66624-AE
AAJ66624-AP
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), liquid for electrophoresis
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Triton® X-100, Electrophoresis Reagent
CAS No.: 9002-93-1
Formula Weight: ca. 625
Flash Point: 180°(356°F)
Physical Form: Liquid
Density: 1.061
Appearance: Corlorless to pale yellow
Odor: Mild
Melting Point: ca. 5°
Boiling Point:
Boiling Pt:
>250 °C Melting Pt: ca. 5 °C Density: 1.061 Flash Pt: 180 °C(356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00132505 CAS Number: 9002-93-1 ADR: 9,III Merck Index: 14,6761 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...