Ceftazidime pentahydrate 95%

Supplier: Thermo Scientific Chemicals
Danger

J66460-03 J66460-06
10067-164EA 141.04 USD
10067-164 10067-166
Ceftazidime pentahydrate 95%
Ceftazidime pentahydrate

Targets primarily Gram-negative bacteria especially Pseudomonas aeruginosa unlike other third generation cephalosporins


Purity: 95% CAS No.: 78439-06-2 Molecular Formula: C22H22N6O7S2.5H2O Formula Weight: 636.65 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Solubility: Sparingly soluble in aqueous solution MDL No.: MFCD00153936


Applications: Targets primarily gram negative bacteria especially Pseudomonas aeruginosa unlike other third generation cephalosporins

Formula: C₂₂H₃₂N₆O₁₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00153936
CAS Number: 78439-06-2

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR