Piperacillin sodium salt 835-1007 µg/mg

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

J66419-06 J66419-09 J66419-03
10067-148EA 316 USD
10067-148 10067-150 AAJ66419-03
Piperacillin sodium salt 835-1007 µg/mg
Piperacillin sodium salt

A broad spectrum antibiotic targeting a wide range of susceptible Gram-positive and Gram-negative bacteria


Applications: A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria


Applications: A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria

Formula: C₂₃H₂₇N₅O₇S
MW: 539.54 g/mol
Boiling Pt: 793 °C
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00056867
CAS Number: 59703-84-3
EINECS: 261-868-6
Merck Index: 14,7463

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR