Itraconazole 98%

Supplier: Thermo Scientific Chemicals
Warning

J66390-06 J66390-14 J66390-03
10067-140EA 180 USD
10067-140 10067-142 AAJ66390-03
Itraconazole 98%
Itraconazole

Itraconazole, 98%


Purity: 98% CAS No.: 84625-61-6 Molecular Formula: C35H38Cl2N8O4 Formula Weight: 705.63 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 166° Solubility: Insoluble in water. Solube in chloroform at 50 mg/ml. Slightly soluble in ethanol or methanol MDL No.: MFCD00870168

Formula: C₃₅H₃₈Cl₂N₈O₄
Melting Pt: 166 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00870168
CAS Number: 84625-61-6
Merck Index: 14,5245

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR